MilliporeSigma
  • Are extraction methods in quantitative assays of pharmacopoeia monographs exhaustive? A comparison with pressurized liquid extraction.
Are extraction methods in quantitative assays of pharmacopoeia monographs exhaustive? A comparison with pressurized liquid extraction.
Planta medica (2006-08-23)
Carlos Basalo, Tobias Mohn, Matthias Hamburger
ABSTRACT
The extraction methods in selected monographs of the European and the Swiss Pharmacopoeia were compared to pressurized liquid extraction (PLE) with respect to the yield of constituents to be dosed in the quantitative assay for the respective herbal drugs. The study included five drugs, Belladonnae folium, Colae semen, Boldo folium, Tanaceti herba and Agni casti fructus. They were selected to cover different classes of compounds to be analyzed and different extraction methods to be used according to the monographs. Extraction protocols for PLE were optimized by varying the solvents and number of extraction cycles. In PLE, yields > 97 % of extractable analytes were typically achieved with two extraction cycles. For alkaloid-containing drugs, the addition of ammonia prior to extraction significantly increased the yield and reduced the number of extraction cycles required for exhaustive extraction. PLE was in all cases superior to the extraction protocol of the pharmacopoeia monographs (taken as 100 %), with differences ranging from 108 % in case of parthenolide in Tanaceti herba to 343 % in case of alkaloids in Boldo folium.
Product Description
Supelco
Boldine, analytical standard