MilliporeSigma
  • Spectral studies on Co(II), Ni(II) and Cu(II) complexes with thiosemicarbazone (L1) and semicarbazone (L2) derived from 2-acetyl furan.
Spectral studies on Co(II), Ni(II) and Cu(II) complexes with thiosemicarbazone (L1) and semicarbazone (L2) derived from 2-acetyl furan.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2006-08-22)
Sulekh Chandra, Anil Kumar
ABSTRACT
Co(II), Ni(II) and Cu(II) complexes are synthesized with thiosemicarbazone (L1) and semicarbazone (L2) derived from 2-acetyl furan. These complexes are characterized by elemental analysis, molar conductance, magnetic susceptibility measurements, mass, IR, electronic and EPR spectral studies. The molar conductance measurements of the complexes in DMSO correspond to non-electrolytic nature except Ni(L)2(NO3)2, which is 1:2 electrolyte. All the complexes are of high-spin type. On the basis of spectral studies an octahedral geometry may be assigned for Co(II) and Ni(II) complexes except nitrato complexes of Ni(II) which is of tetrahedral geometry, whereas tetragonal geometry for Cu(II) complexes.
Sigma-Aldrich
2-Furyl methyl ketone, ≥99%, FG
Sigma-Aldrich
2-Furyl methyl ketone, 99%
Sigma-Aldrich
2-Furyl methyl ketone, natural (US), ≥97%, FG