MilliporeSigma
  • Simple synthesis and anti-HIV activity of novel 3'-vinyl branched apiosyl pyrimidine nucleosides.
Simple synthesis and anti-HIV activity of novel 3'-vinyl branched apiosyl pyrimidine nucleosides.
Nucleosides, nucleotides & nucleic acids (2006-08-12)
Chang Hyun Oh, Jin Woo Kim, Joon Hee Hong
ABSTRACT
Novel vinyl branched apiosyl nucleosides were synthesized in this study. Apiosyl sugar moiety was constructed by sequential ozonolysis and reductions. The bases (uracil and thymine) were efficiently coupled by glycosyl condensation procedure (persilyated base and TMSOTf). The antiviral activities of the synthesized compounds were evaluated against the HIV-1, HSV-1, HSV-2, and HCMV. Compound 10beta displayed moderate anti-HIV activity (EC50 = 17.3 microg/mL) without exhibiting any cytotoxicity up to 100 microM.