MilliporeSigma
  • A codon deletion confers resistance to herbicides inhibiting protoporphyrinogen oxidase.
A codon deletion confers resistance to herbicides inhibiting protoporphyrinogen oxidase.
Proceedings of the National Academy of Sciences of the United States of America (2006-08-09)
William L Patzoldt, Aaron G Hager, Joel S McCormick, Patrick J Tranel
ABSTRACT
Herbicides that act by inhibiting protoporphyrinogen oxidase (PPO) are widely used to control weeds in a variety of crops. The first weed to evolve resistance to PPO-inhibiting herbicides was Amaranthus tuberculatus, a problematic weed in the midwestern United States that previously had evolved multiple resistances to herbicides inhibiting two other target sites. Evaluation of a PPO-inhibitor-resistant A. tuberculatus biotype revealed that resistance was a (incompletely) dominant trait conferred by a single, nuclear gene. Three genes predicted to encode PPO were identified in A. tuberculatus. One gene from the resistant biotype, designated PPX2L, contained a codon deletion that was shown to confer resistance by complementation of a hemG mutant strain of Escherichia coli grown in the presence and absence of the PPO inhibitor lactofen. PPX2L is predicted to encode both plastid- and mitochondria-targeted PPO isoforms, allowing a mutation in a single gene to confer resistance to two herbicide target sites. Unique aspects of the resistance mechanism include an amino acid deletion, rather than a substitution, and the dual-targeting nature of the gene, which may explain why resistance to PPO inhibitors has been rare.
Product Description
Supelco
Lactofen, PESTANAL®, analytical standard