MilliporeSigma
  • Structural basis for the design of potent and species-specific inhibitors of 3-hydroxy-3-methylglutaryl CoA synthases.
Structural basis for the design of potent and species-specific inhibitors of 3-hydroxy-3-methylglutaryl CoA synthases.
Proceedings of the National Academy of Sciences of the United States of America (2006-07-26)
Florence Pojer, Jean-Luc Ferrer, Stéphane B Richard, Dinesh A Nagegowda, Mee-Len Chye, Thomas J Bach, Joseph P Noel
ABSTRACT
3-Hydroxy-3-methylglutaryl CoA synthase (HMGS) catalyzes the first committed step in the mevalonate metabolic pathway for isoprenoid biosynthesis and serves as an alternative target for cholesterol-lowering and antibiotic drugs. We have determined a previously undescribed crystal structure of a eukaryotic HMGS bound covalently to a potent and specific inhibitor F-244 [(E,E)-11-[3-(hydroxymethyl)-4-oxo-2-oxytanyl]-3,5,7-trimethyl-2,4-undecadienenoic acid]. Given the accessibility of synthetic analogs of the F-244 natural product, this inhibited eukaryotic HMGS structure serves as a necessary starting point for structure-based methods that may improve the potency and species-specific selectivity of the next generation of F-244 analogs designed to target particular eukaryotic and prokaryotic HMGS.
Sigma-Aldrich
Hymeglusin, ≥98% (HPLC), from Fusarium sp.