MilliporeSigma
  • Subcellular localization and possible functions of gamma-glutamyltransferase in the radish (Raphanus sativus L.) plant.
Subcellular localization and possible functions of gamma-glutamyltransferase in the radish (Raphanus sativus L.) plant.
Bioscience, biotechnology, and biochemistry (2006-07-25)
Yoshihiro Nakano, Satoshi Okawa, Rafael Prieto, Jiro Sekiya
ABSTRACT
Previously we reported the purification of soluble gamma-glutamyltransferases (GGTs) from radish cotyledon. Subcellular fractionation of radish cells revealed that soluble GGT is a vacuolar enzyme. Acivicin, a GGT inhibitor, mediated the in vivo catabolism inhibition of the glutathione S-conjugate generated from endogenous glutathione and exogenously supplied monochlorobimane. Thus soluble GGT is possibly involved in the catabolism of glutathione S-conjugates.
Sigma-Aldrich
Monochlorobimane, suitable for fluorescence, ≥70.0% (HPCE)