MilliporeSigma
  • 15N spin diffusion rate in solid-state NMR of totally enriched proteins: the magic angle spinning frequency effect.
15N spin diffusion rate in solid-state NMR of totally enriched proteins: the magic angle spinning frequency effect.
Journal of magnetic resonance (San Diego, Calif. : 1997) (2006-07-21)
Alexey Krushelnitsky, Thomas Bräuniger, Detlef Reichert
ABSTRACT
As demonstrated by means of the one-dimensional solid-state MAS exchange experiment (CODEX), the rate of the proton driven spin diffusion between backbone (15)N nuclei in totally enriched protein depends strongly on the magic angle spinning (MAS) frequency: spin diffusion at MAS frequency 16 kHz is about 4-5 times slower as compared to that at MAS frequency 1 kHz which is due to the averaging of the homo- and hetero-nuclear dipolar interactions by MAS. It is important that even at the highest MAS frequencies used in our experiments the spin diffusion rate is comparable or larger than typical values of the spin-lattice relaxation rates of backbone nitrogens in solid proteins. Thus, the precise quantitative analysis of (15)N T(1)'s in totally enriched solid proteins may lead to wrong quantitative results. On the other hand, the effectiveness of the (15)N-(15)N correlation and structure determination experiments making use of (15)N-(15)N distances can be increased by decreasing the MAS frequency as far as possible, which is counter intuitive to the commonly applied fast MAS conditions in order to reduce the dipolar-broadened line widths for increased spectral resolution.
Sigma-Aldrich
Boc-Gly-OH, ≥99.0% (T)
Pricing and availability is not currently available.