MilliporeSigma
NO2 adsorption on BaO/Al2O3: the nature of nitrate species.
The journal of physical chemistry. B (2006-07-21)
Janós Szanyi, Ja Hun Kwak, Do Heui Kim, Sarah D Burton, Charles H F Peden
ABSTRACT
Temperature programmed desorption, infrared spectroscopy, and (15)N solid state NMR spectroscopy were used to characterize the nature of the nitrate species formed on Al(2)O(3) and BaO/Al(2)O(3) NO(x) storage/reduction materials. Two distinctly different nitrate species were found: surface nitrates that are associated with a monolayer BaO on the alumina support, and a bulk-like nitrate that forms on this thin BaO layer. The surface nitrates desorb as NO(2) at lower temperatures than do the bulk-like nitrates, which decompose as NO+O(2) at higher temperatures. The amount of NO(x) stored in the monolayer nitrate is proportional to the surface area of the catalyst, while that in the bulk nitrate increases with BaO coverage.
Sigma-Aldrich
Barium oxide, 99.99% trace metals basis
Sigma-Aldrich
Barium oxide, 97%