MilliporeSigma
  • Effects of penconazole on two yeast strains: growth kinetics and molecular studies.
Effects of penconazole on two yeast strains: growth kinetics and molecular studies.
Molecular nutrition & food research (2006-05-27)
Dalal Jawich, Roger Lteif, Annie Pfohl-Leszkowicz, Pierre Strehaiano
ABSTRACT
The aim of this study consisted to evaluate the impact of a pesticide (penconazole) on the growth kinetics and genotoxicity on two yeast strains (Saccharomyces cerevisiae and Metschnikowia pulcherrima). When the penconazole was added at different phases of the growth of M. pulcherrima, no effect was noticed on the kinetics of yeast growth but DNA adducts were observed when penconazole was added in the exponential phase. Increasing doses (1-15 maximum residue limit) of the pesticide added at the beginning of the fermentation did not induce DNA adducts while kinetics were affected.
Product Description
Supelco
Penconazol, PESTANAL®, analytical standard