MilliporeSigma
  • Interception of the radicals produced in electrophilic fluorination with radical traps (Tempo, Dmpo) studied by electrospray ionization mass spectrometry.
Interception of the radicals produced in electrophilic fluorination with radical traps (Tempo, Dmpo) studied by electrospray ionization mass spectrometry.
Rapid communications in mass spectrometry : RCM (2006-05-24)
Xiang Zhang, Haoyang Wang, Yinlong Guo
ABSTRACT
The interaction of the nitroxide radical traps (Tempo and Dmpo) and radicals produced in the electrophilic fluorination of olefins (styrene and alpha-methylstyrene) and Selectfluor (1-chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octanebis(tetrafluoroborate) (F-TEDA-BF(4)) (1)) was investigated by electrospray ionization mass spectrometry (ESI-MS). Tempo succeeded in intercepting the radical cationic intermediates and the radical adduct ions were detected at m/z 260 (for styrene) and m/z 274 (for alpha-methylstyrene). Dmpo could also intercept the fluorine radical and radical adduct ions were detected at m/z 131, 132 and 152. The interception of the radical cationic intermediates and fluorine radical is good evidence for the presence of a single-electron transfer mechanism in the electrophilic fluorination.
Sigma-Aldrich
Methylstyrene, 60% meta, 40% para and 1% ortho, 99%, contains ~50 ppm 4-tert-butylcatechol as inhibitor