MilliporeSigma
  • Superoxide-dependent reduction of some simple low molecular mass iron complexes.
Superoxide-dependent reduction of some simple low molecular mass iron complexes.
Journal of trace elements and electrolytes in health and disease (1991-12-01)
J M Gutteridge
ABSTRACT
Superoxide and hydrogen peroxide are often ascribed iron reducing roles in reactions leading to the formation of an aggressive oxidant by Fenton Chemistry. Hydrogen peroxide, however, does not appear to reduce iron in Fenton reactions, and we here show that several simple ferric complexes are not readily reduced to the ferrous state by superoxide generating systems. Our knowledge of low molecular mass iron complexes in vivo and their reactivity with superoxide radicals is at present insufficient for much of the chemistry proposed.
Sigma-Aldrich
3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5′,5′′-disulfonic acid disodium salt, Suitable for determination of Fe
Supelco
3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5′,5′′-disulfonic acid disodium salt, for spectrophotometric det. of Fe, ≥99.0%