MilliporeSigma
A comparative study of five fatal cases of Taxus poisoning.
International journal of legal medicine (2006-05-09)
J Pietsch, K Schulz, U Schmidt, H Andresen, B Schwarze, J Dressler
ABSTRACT
The study presents five fatal cases of poisoning with Taxus spp., all of which were suicides of young people aged between 16 and 26 years. Yew leaves were consumed in four fatalities, whereas a mash from Taxus was ingested in one case. No relevant concentrations of alcohol, narcotic drugs, and pharmaceuticals were determined in postmortem toxicological screening. At forensic autopsy, a widely dilated pupil was found in two decedents. Furthermore, autopsy showed unspecific findings of intoxication in all cases: acute blood congestion of lungs, liver, kidney, and brain as well as dilated cardiac ventricles. No signs of violence could be found in any of the fatalities. Yew leaves were identified in four cases in the stomach and duodenum. 3,5-Dimethoxyphenol, the aglycon of the Taxus ingredient taxicatine, was determined as toxicological evidence for the absorption of yew ingredients. Taxus intoxication could be confirmed by 3,5-dimethoxyphenol concentrations in cardiac blood between 31 and 528 ng/ml for all cases. 3,5-Dimethoxyphenol was also detected in stomach contents as well as in urine, liver, kidneys, and brain samples. Based on the different concentrations of 3,5-dimethoxyphenol determined in the cardiac blood samples, it was concluded that the form of ingestion plays a decisive role in the process of poisoning. Finally, a toxic range for Taxus poisoning based on 3,5-dimethoxyphenol as marker substance is proposed as orientation.
Sigma-Aldrich
3,5-Dimethoxyphenol, 99%