MilliporeSigma
  • Aziridinium from N,N-dibenzyl serine methyl ester: synthesis of enantiomerically pure beta-amino and alpha,beta-diamino esters.
Aziridinium from N,N-dibenzyl serine methyl ester: synthesis of enantiomerically pure beta-amino and alpha,beta-diamino esters.
Organic letters (2006-05-05)
Cédric Couturier, Jérome Blanchet, Thierry Schlama, Jieping Zhu
ABSTRACT
[reaction: see text] Reaction of N,N-dibenzyl-O-methylsulfonyl serine methyl ester with a variety of heteronucleophiles (sodium azide, sodium phthalimide, amines, thiols) and carbanions (sodium malonate) gave, via an aziridinium intermediate, the corresponding beta-amino or alpha,beta-diamino ester in good to excellent yield. A short synthesis of orthogonally protected and enantiomerically pure 2,3-diamino propionate (Dap) is described.
Sigma-Aldrich
L-Serine methyl ester hydrochloride, 98%
Sigma-Aldrich
DL-Serine methyl ester hydrochloride, 98%
Sigma-Aldrich
D-Serine methyl ester hydrochloride, 98%