MilliporeSigma
  • Delivery of hydrophobised 5-fluorouracil derivative to brain tissue through intravenous route using surface modified nanogels.
Delivery of hydrophobised 5-fluorouracil derivative to brain tissue through intravenous route using surface modified nanogels.
Journal of drug targeting (2006-04-13)
Sheetal Soni, Anil K Babbar, Rakesh K Sharma, Amarnath Maitra
ABSTRACT
Random copolymeric micelles composed of N-isopropylacrylamide (NIPAAM) and N-vinylpyrrolidone (VP) cross-linked with N,N'-methylenebisacrylamide (MBA) have been used as nanogel carriers to encapsulate N-hexylcarbamoyl-5-fluorouracil (HCFU), a prodrug of 5-FU, and have been targeted to brain tissue across blood-brain barrier (BBB) after coating with polysorbate 80. Accumulation of nanogel particles in the brain and other tissues of "strain A" mice had been monitored by radiolabeling of nanogels with (99m)Tc. Gamma Scintigraphic technique was also performed to see the distribution of (99m)Tc labeled nanogels in the brain. The retention time in blood appeared to be slightly longer for coated nanogels than that of uncoated nanogels though the accumulation of coated nanogels in the RES was more or less same as that of uncoated nanogels. The blood however had almost double accumulation of polysorbate 80 coated nanogels in the initial 5 min compared to that shown by uncoated nanogels. We speculate that coating of nanogels with polysorbate 80 alters the surface properties of nanogels, which results in relatively higher uptake in the brain tissue. The studies revealed that a large portion of (99m)Tc labeled HCFU loaded nanogels are accumulated in the RES (lung, liver and spleen). The accumulation of the labeled nanogels in the brain, however, is much less compared to RES and it has been found that while an amount of uncoated labeled nanogels was found to be 0.18% of the injected dose, it increased to 0.52% on coating with polysorbate 80. The optimal amount of polysorbate 80 added to nanogels for the maximum delivery of particles to brain was found to be 1% w/w. These results were further supported by the gamma scintigrams of New Zealand rabbits. Thus, the present nanogel system has opened a new avenue for poorly soluble drugs to be targeted to brain by coating the particles with polysorbate 80.
Sigma-Aldrich
Carmofur, ≥98% (HPLC), powder