MilliporeSigma
  • Simultaneous spectrophotometric determination of paracetamol and salicylamide in human serum and pharmaceutical formulations by a differential kinetic method.
Simultaneous spectrophotometric determination of paracetamol and salicylamide in human serum and pharmaceutical formulations by a differential kinetic method.
Journal of AOAC International (2006-03-11)
Ali Reza Zarei, Abbas Afkhami, Nahid Sarlak
ABSTRACT
A rapid, simple, and sensitive differential kinetic method is presented for the determinations of acetaminophen (also known as paracetamol) and salicylamide. The method is based on their oxidation reaction by Fe3+ ion in the presence of 1, 10-phenanthroline as indicator. The reactions can be monitored spectrophotometrically by measuring the increase in the absorbance of the solution at 510 nm. Two times were selected one in which only paracetamol is oxidized by Fe3+ ion and the other in which both drugs are oxidized by Fe3+ ion. The data were evaluated by the proportional equations method. The method allowed the simultaneous determination of paracetamol and salicylamide at concentrations between 0.5-20 and 1-40 microg/mL with relative standard deviations of 3.47 and 2.58%, respectively. The method was applied to the simultaneous determination of paracetamol and salicylamide in human serum and pharmaceutical formulations.
Sigma-Aldrich
Salicylamide, puriss., ≥99.0% (T)
Sigma-Aldrich
Salicylamide, 99%