MilliporeSigma
  • Laboratory simulated dissipation of metsulfuron methyl and chlorimuron ethyl in soils and their residual fate in rice, wheat and soybean at harvest.
Laboratory simulated dissipation of metsulfuron methyl and chlorimuron ethyl in soils and their residual fate in rice, wheat and soybean at harvest.
Journal of Zhejiang University. Science. B (2006-02-28)
Nilanjan Sanyal, Sukhendu Kumar Pramanik, Raktim Pal, Ashim Chowdhury
ABSTRACT
Two sulfonylurea herbicides, metsulfuron methyl (Ally 20 WP) and chlorimuron ethyl (Classic 25 WP) were evaluated for their dissipation behaviour in alluvial, coastal saline and laterite soils under laboratory incubated condition at 60% water holding capacity of soils and 30 degrees C temperature was maintained. In field study herbicides were applied twice for the control of grasses, annual and perennials broad leaves weeds and sedges in rice, wheat and soybean to find out the residual fate of both the herbicides on different matrices of respective crops after harvest. Extraction and clean up methodologies for the herbicides were standardized and subsequently analyzed by HPLC. The study revealed that the half-lives of metsulfuron methyl and chlorimuron ethyl ranged from 10.75 to 13.94 d irrespective of soils and doses applied. Field trials with rice, wheat and soybean also revealed that these two herbicides could safely be recommended for application as no residues were detected in the harvest samples.
Product Description
Supelco
Chlorimuron ethyl, PESTANAL®, analytical standard