MilliporeSigma
  • Novel synthesis of desymmetrized resorcinol derivatives: aryl fluoride displacement on deactivated substrates.
Novel synthesis of desymmetrized resorcinol derivatives: aryl fluoride displacement on deactivated substrates.
The Journal of organic chemistry (2006-02-25)
Aujin Kim, Jeremiah D Powers, Jennifer F Toczko
ABSTRACT
A short, high-yielding synthesis of differentially substituted resorcinol derivatives has been developed that utilizes 1,3-difluorobenzene as the starting material and employs sequential nucleophilic aromatic substitution (S(N)Ar) reactions to generate desymmetrized products. The scope and limitations of the second S(N)Ar reaction on the deactivated 1-alkoxy-3-fluorobenzene intermediates have been investigated. This methodology has also been employed in the synthesis of desymmetrized catechol derivatives from 1,2-difluorobenzene.
Sigma-Aldrich
1,2-Difluorobenzene, 98%
Sigma-Aldrich
1,3-Difluorobenzene, ≥99%