MilliporeSigma
Nickel-catalyzed coupling of allyl chlorides and enynes.
Chemical communications (Cambridge, England) (2006-02-24)
Shin-ichi Ikeda, Kaori Suzuki, Kazunori Odashima
ABSTRACT
The Ni-catalyzed coupling of allyl chlorides and enynes has been developed; the cyclization of enynes was triggered by the addition of pi-allylnickel species to the alkyne part, followed by the incorporation of the alkene part.
Sigma-Aldrich
Allyl chloride, ReagentPlus®, 99%
Sigma-Aldrich
Allyl chloride, reagent grade, 98%