MilliporeSigma
  • Solution and structural investigations of ligand preorganization in trivalent lanthanide complexes of bicyclic malonamides.
Solution and structural investigations of ligand preorganization in trivalent lanthanide complexes of bicyclic malonamides.
Inorganic chemistry (2006-02-14)
Bevin W Parks, Robert D Gilbertson, James E Hutchison, Elisabeth Rather Healey, Timothy J R Weakley, Brian M Rapko, Benjamin P Hay, Sergei I Sinkov, Grant A Broker, Robin D Rogers
ABSTRACT
This report describes an investigation into the coordination chemistry of trivalent lanthanides in solution and the solid state with acyclic and preorganized bicyclic malonamide ligands. Two experimental investigations were performed: solution binding affinities were determined through single-phase spectrophotometric titrations and the extent of conformational change upon binding was investigated with single-crystal X-ray crystallography. Both experimental methods compare the bicyclic malonamide (BMA), which is designed to be preorganized for binding trivalent lanthanides, to an analogous acyclic malonamide. Results from the spectrophotometric titrations indicate that BMA exhibits a 10-100x increase in binding affinity to Ln(III) over acyclic malonamide. In addition, BMA forms compounds with high ligand-metal ratios, even when competing with water and nitrate ligands for binding sites. The crystal structures exhibit no significant differences in the nature of the binding between Ln(III) and the BMA or acyclic malonamide. These results support the conclusion that rational ligand design can lead to compounds that enhance the binding affinities within a ligand class.
Sigma-Aldrich
Malonamide, 97%