MilliporeSigma
  • Thermodynamics of interactions of water-soluble porphyrins with RNA duplexes.
Thermodynamics of interactions of water-soluble porphyrins with RNA duplexes.
Journal of the American Chemical Society (2006-02-09)
Ara A Ghazaryan, Yeva B Dalyan, Samvel G Haroutiunian, Anna Tikhomirova, Nicolas Taulier, James W Wells, Tigran V Chalikian
ABSTRACT
We characterized the interactions of meso-tetrakis(4N-(2-hydroxyethyl)pyridinium-4-yl) porphyrin (TEtOHPyP4), meso-tetrakis(4N-allylpyridinium-4-yl) porphyrin (TAlPyP4), and meso-tetrakis(4N-metallylpyridinium-4-yl) porphyrin (TMetAlPyP4) with the poly(rA)poly(rU) and poly(rI)poly(rC) RNA duplexes between 18 and 45 degrees C by employing circular dichroism, light absorption, and fluorescence intensity spectroscopic measurements. Our results suggest that TEtOHPyP4 and TAlPyP4 intercalate into the poly(rA)poly(rU) and poly(rI)poly(rC) host duplexes, while TMetAlPyP4 associates with these RNA duplexes by forming outside-bound, self-stacked aggregates. We used our temperature-dependent absorption titration data to determine the binding constants and stoichiometry for each porphyrin-RNA binding event studied in this work. From the temperature dependences of the binding constants, we calculated the binding free energies, DeltaG(b), enthalpies, DeltaH(b), and entropies, DeltaS(b). For each RNA duplex, the binding enthalpy, DeltaH(b), is the most favorable for TEtOHPyP4 (an intercalator) followed by TAlPyP4 (an intercalator) and TMetAlPyP4 (an outside binder). On the other hand, for each duplex, external self-stacking of TMetAlPyP4 produces the most favorable change in entropy, DeltaS(b), followed by the intercalators TAlPyP4 and TEtOHPyP4. Thus, our results suggest that the thermodynamic profile of porphyrin-RNA binding may correlate with the binding mode. This correlation reflects the differential nature of molecular forces that stabilize/destabilize the two modes of binding-intercalation versus external self-stacking along the host duplex.
Sigma-Aldrich
Polyadenylic acid-Polyuridylic acid sodium salt, double-stranded homopolymer