MilliporeSigma
  • Bothrops moojeni myotoxin-II, a Lys49-phospholipase A2 homologue: an example of function versatility of snake venom proteins.
Bothrops moojeni myotoxin-II, a Lys49-phospholipase A2 homologue: an example of function versatility of snake venom proteins.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP (2006-01-31)
Rodrigo G Stábeli, Saulo F Amui, Carolina D Sant'Ana, Matheus G Pires, Auro Nomizo, Marta C Monteiro, Pedro R T Romão, Renata Guerra-Sá, Carlos A Vieira, José R Giglio, Marcos R M Fontes, Andreimar M Soares
ABSTRACT
MjTX-II, a myotoxic phospholipase A(2) (PLA(2)) homologue from Bothrops moojeni venom, was functionally and structurally characterized. The MjTX-II characterization included: (i) functional characterization (antitumoral, antimicrobial and antiparasitic effects); (ii) effects of structural modifications by 4-bromophenacyl bromide (BPB), cyanogen bromide (CNBr), acetic anhydride and 2-nitrobenzenesulphonyl fluoride (NBSF); (iii) enzymatic characterization: inhibition by low molecular weight heparin and EDTA; and (iv) molecular characterization: cDNA sequence and molecular structure prediction. The results demonstrated that MjTX-II displayed antimicrobial activity by growth inhibition against Escherichia coli and Candida albicans, antitumoral activity against Erlich ascitic tumor (EAT), human breast adenocarcinoma (SK-BR-3) and human T leukemia cells (JURKAT) and antiparasitic effects against Schistosoma mansoni and Leishmania spp., which makes MjTX-II a promising molecular model for future therapeutic applications, as well as other multifunctional homologous Lys49-PLA(2)s or even derived peptides. This work provides useful insights into the structural determinants of the action of Lys49-PLA(2) homologues and, together with additional strategies, supports the concept of the presence of others "bioactive sites" distinct from the catalytic site in snake venom myotoxic PLA(2)s.
Product Description
Supelco
2,4′-Dibromoacetophenone, for HPLC derivatization, LiChropur, ≥99.0% (HPLC)
Sigma-Aldrich
2,4′-Dibromoacetophenone, >98%