MilliporeSigma
  • Sulfation and glucuronidation of phenols: implications in coenyzme Q metabolism.
Sulfation and glucuronidation of phenols: implications in coenyzme Q metabolism.
Methods in enzymology (2006-01-10)
Nandita Shangari, Tom S Chan, Peter J O'Brien
ABSTRACT
Phase II conjugation of phenolic compounds constitutes an important mechanism through which exogenous or endogenous toxins are detoxified and excreted. Species differences in the rates of glucuronidation or sulfation can lead to significant variation in the metabolism of this class of compounds. Conjugation of the hydroxyl groups of phenols can occur with glucuronate or sulfate. Quinone metabolism, deactivation, and detoxification are also affected by the same conjugatory systems as phenols; however, reduction of quinones to hydroquinols seems to be a prerequisite. This work reviews current knowledge on phenol conjugation and its implications on hydroquinone metabolism with special consideration for coenzyme Q metabolism.
Sigma-Aldrich
Coenzyme Q1, ≥95%