MilliporeSigma
  • Sodium dichloroisocyanurate (NaDCC) tablets as an alternative to sodium hypochlorite for the routine treatment of drinking water at the household level.
Sodium dichloroisocyanurate (NaDCC) tablets as an alternative to sodium hypochlorite for the routine treatment of drinking water at the household level.
International journal of hygiene and environmental health (2006-01-03)
Thomas Clasen, Paul Edmondson
ABSTRACT
Household water treatment using sodium hypochlorite (NaOCl) has been recognized as a cost-effective means of reducing the heavy burden of diarrhea and other waterborne diseases, especially among populations without access to improved water supplies. Sodium dichloroisocyanurate (NaDCC), which is widely used in emergencies, is an alternative source of chlorine that may present certain advantages over NaOCl for household-based interventions in development settings. We summarize the basic chemistry and possible benefits of NaDCC, and review the available literature concerning its safety and regulatory treatment and microbiological effectiveness. We review the evidence concerning NaDCC in field studies, including microbiological performance and health outcomes. Finally, we examine studies and data to compare NaDCC with NaOCl in terms of compliance, acceptability, affordability and sustainability, and suggest areas for further research.
Sigma-Aldrich
Sodium dichloroisocyanurate, 96%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dichloroisocyanurate dihydrate, 98.0% (AT)
Pricing and availability is not currently available.