MilliporeSigma
  • Sodium stibogluconate resistance in Leishmania donovani correlates with greater tolerance to macrophage antileishmanial responses and trivalent antimony therapy.
Sodium stibogluconate resistance in Leishmania donovani correlates with greater tolerance to macrophage antileishmanial responses and trivalent antimony therapy.
Parasitology (2005-12-13)
K C Carter, S Hutchison, A Boitelle, H W Murray, S Sundar, A B Mullen
ABSTRACT
Co-treatment of mice infected with different strains of Leishmania donovani with a non-ionic surfactant vesicle formulation of buthionine sulfoximine (BSO-NIV), and sodium stibogluconate (SSG), did not alter indicators of Th1 or Th2 responses but did result in a significant strain-independent up-regulation of IL6 and nitrite levels by stimulated splenocytes from treated mice compared to controls. The efficacy of BSO-NIV/SSG treatment was dependent on the host being able to mount a respiratory burst indicating that macrophages are important in controlling the outcome of treatment. In vitro studies showed that SSG resistance was associated with a greater resistance to killing by activated macrophages, treatment with hydrogen peroxide or potassium antimony tartrate. Longitudinal studies showed that a SSG resistant (SSG-R) strain was more virulent than a SSG susceptible (SSG-S) strain, resulting in significantly higher parasite burdens by 4 months post-infection. These results indicate that SSG exposure may favour the emergence of more virulent strains.
Sigma-Aldrich
Potassium antimonyl tartrate trihydrate, puriss., meets analytical specification of USP, 99.0-103.0%, powder
Sigma-Aldrich
Potassium antimonyl tartrate trihydrate, purum p.a., 99.0-103% (RT)
Sigma-Aldrich
Potassium antimonyl tartrate trihydrate, ACS reagent, ≥99%