MilliporeSigma
  • On-line continuous potentiometric measurement of potassium concentration in whole blood during open-heart surgery.
On-line continuous potentiometric measurement of potassium concentration in whole blood during open-heart surgery.
Clinical chemistry (1979-01-01)
H F Osswald, R Asper, W Dimai, W Simon
PMID761376
ABSTRACT
We describe a flow-through system with an ion-selective electrode for measurement of blood potassium ion concentration, continuously and on-line off the extracorporeal blood circulation in an operating theater during human open-heart surgery. Comparison measurements were made with the SMA flame photometer (blood plasma) and an Orion SS 30 sodium/potassium analyzer (whole blood). The potassium concentration values obtained with the flow-through system agree well with the ones determined with the flame photometer. The time delay of the measurement with the flow-through system was relatively long (2 min) but delays of only 10--20 s seem feasible. Short time delays can deepen insight and simplify rational treatment under surgery conditions.
Product Description
Supelco
Potassium ionophore I, Selectophore, for ion-selective electrodes, function tested
Pricing and availability is not currently available.