MilliporeSigma
  • In vitro activity of lauric acid or myristylamine in combination with six antimicrobial agents against methicillin-resistant Staphylococcus aureus (MRSA).
In vitro activity of lauric acid or myristylamine in combination with six antimicrobial agents against methicillin-resistant Staphylococcus aureus (MRSA).
International journal of antimicrobial agents (2005-12-02)
Takashi Kitahara, Yuko Aoyama, Yoichi Hirakata, Shimeru Kamihira, Shigeru Kohno, Nobuhiro Ichikawa, Mikiro Nakashima, Hitoshi Sasaki, Shun Higuchi
ABSTRACT
The objective of this study was to investigate the in vitro activities of lauric acid and myristylamine in combination with six antimicrobial agents against methicillin-resistant Staphylococcus aureus (MRSA). The combination effect of lipids and antimicrobial agents was evaluated by the checkerboard method to obtain a fractional inhibitory concentration (FIC) index. The effects of lauric acid + gentamicin (GM) and lauric acid + imipenem (IPM) combinations were synergistic against the clinical isolates in 12 combinations. An antagonistic FIC index was observed only with the myristylamine + GM combination. We investigated in detail the antimicrobial activity for two combinations that showed a synergistic effect. The cytotoxicity of lauric acid was not enhanced by the addition of GM and IPM. In time-kill studies, lauric acid + GM and lauric acid + IPM combinations at one-eighth of the minimum inhibitory concentration produced a bacteriostatic effect.
Sigma-Aldrich
Tetradecylamine, 95%