MilliporeSigma
  • Roquefortine E, a diketopiperazine from an Australian isolate of Gymnoascus reessii.
Roquefortine E, a diketopiperazine from an Australian isolate of Gymnoascus reessii.
Journal of natural products (2005-11-29)
Ben Clark, Robert J Capon, Ernest Lacey, Shaun Tennant, Jennifer H Gill
ABSTRACT
The new isoprenylated diketopiperazine roquefortine E (6) has been isolated from an Australian soil isolate of the ascomycete Gymnoascus reessii. The known fungal metabolite roquefortine C (1) was also recovered as the major antibacterial principle, and all structures were assigned by detailed spectroscopic analysis.
Sigma-Aldrich
Roquefortine C, ≥98% (HPLC), from Penicillium roqueforti