MilliporeSigma
  • Allopurinol suppresses 2-bromoethylamine and 1-methyl-4-phenylpyridinium ion (MPP(+))-induced hydroxyl radical generation in rat striatum.
Allopurinol suppresses 2-bromoethylamine and 1-methyl-4-phenylpyridinium ion (MPP(+))-induced hydroxyl radical generation in rat striatum.
Toxicology (2005-11-08)
Toshio Obata
ABSTRACT
The present study was examined whether or not 2-bromoethyamine, a semicarbazide-sensitive amine oxidase (SSAO, EC; 1.4.3.6) inhibitor, would increase an active dopaminergic neurotoxin, 1-methyl-4-phenylpyridinium ion (MPP(+))-induced hydroxyl radical ((*)OH) generation in the rat striatum. Rats were anesthetized, and sodium salicylate (0.5 mM or 0.5 nmol/microl/min) was infused through a microdialysis probe to detect the generation of (*)OH as reflected by the non-enzymatic formation of 2,3-dihydroxybenzoic acid (DHBA) in the striatum. Infusion of 2-bromoethylamine (100 microM or 100 pmol/microl/min) into the striatum drastically increased the formation of (*)OH products, trapped as DHBA by the action of MPP(+). Further, I studied the effect of allopurinol, a xanthine oxidase inhibitor, an 2-bromoethylamine and MPP(+)-induced (*)OH generation. Allopurinol (10 microM or 10 pmol/microl/min) significantly suppressed 2-bromoethyamine and MPP(+)-induced (*)OH. These results suggest that a definite mechanism is not clear at the moment, after inhibition of tissue-bound and/or blood plasma SSAO activity, with consequent increases in bioactive amine levels, enhances the formation of (*)OH products of efflux/oxidation due to MPP(+).
Sigma-Aldrich
2-Bromoethylamine hydrobromide, purum, ≥97.0% (AT)
Sigma-Aldrich
2-Bromoethylamine hydrobromide, 99%