MilliporeSigma
  • Direct determination of glucuronide and sulfate of p-hydroxymethamphetamine in methamphetamine users' urine.
Direct determination of glucuronide and sulfate of p-hydroxymethamphetamine in methamphetamine users' urine.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2005-11-03)
Noriaki Shima, Hiroe Tsutsumi, Tooru Kamata, Mayumi Nishikawa, Munehiro Katagi, Akihiro Miki, Hitoshi Tsuchihashi
ABSTRACT
Two conjugates of p-hydroxymethamphetamine (p-OHMA), p-OHMA-glucuronide (p-OHMA-Glu) and p-OHMA-sulfate (p-OHMA-Sul) have been identified in methamphetamine (MA) users' urine by using liquid chromatography-high resolution tandem mass spectrometry (LC-HRMS-MS). The synthesis of p-OHMA-Glu and p-OHMA-Sul, and an LC-MS procedure for the simultaneous determination of MA and its four metabolites, amphetamine (AP), p-OHMA, p-OHMA-Glu and p-OHMA-Sul, in urine have also been established. After deproteinizing urine samples with methanol, LC-MS employing a C(18) semi-micro column with a gradient elution program provided the successful separations and MS determinations of these analytes within 20 min. Based on the established method, p-OHMA-Sul was detected at higher concentrations than p-OHMA-Glu in all of the three urine samples tested. These data suggest that sulfation is a major pathway in the MA phase II metabolism.