MilliporeSigma
  • Genotoxic damage in female residents exposed to environmental air pollution in Shenyang city, China.
Genotoxic damage in female residents exposed to environmental air pollution in Shenyang city, China.
Cancer letters (2005-10-26)
Hitoshi Ishikawa, Ying Tian, Fengyuan Piao, Zhiwen Duan, Yumin Zhang, Mingyue Ma, Haishan Li, Hidetaka Yamamoto, Yutaka Matsumoto, Shigekatsu Sakai, Jinshan Cui, Toru Yamauchi, Kazuhito Yokoyama
ABSTRACT
Air pollution has been suggested to cause genetic damage from investigations of many biological markers that measure cytogenetic damage in humans. Here, we evaluated the genotoxic effects of ambient air pollution by investigating the extent of cytogenetic damage in human blood lymphocytes from rural and industrial female residents of Shenyang city, China, using micronuclei assays and polymorphic analyses of metabolic enzyme and DNA repair genes. After adjustment for potential confounding factors including DNA polymorphisms, industrial female residents were found to have a higher micronuclei frequency. These results provide evidence that micronuclei assays are a sensitive indicator to air pollution-induced genotoxic effects in humans.
Product Description
Supelco
Benzo[b]fluoranthene, analytical standard
Sigma-Aldrich
Benzo[b]fluoranthene, 98%
Supelco
Benzo[b]fluoranthene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland