MilliporeSigma
  • Gas-phase chemistry of (alpha-terpineol with ozone and OH radical: rate constants and products.
Gas-phase chemistry of (alpha-terpineol with ozone and OH radical: rate constants and products.
Environmental science & technology (2005-10-06)
J R Wells
ABSTRACT
A bimolecular rate constant, kOH+alpha-terpineol, of (1.9 +/- 0.5) x 10(-10) cm3 molecule(-1) s(-1) was measured using gas chromatography/mass spectrometry and the relative rate technique for the reaction of the hydroxyl radical (OH) with alpha-terpineol (1-methyl-4-isopropyl-1-cyclohexen-8-ol) at (297 +/- 3) K and 1 atm total pressure. Additionally, a bimolecular rate constant, kO3+alpha-terpineol, of (3.0 +/- 0.2) x 10(-16) cm3 molecule(-1) s(-1) was measured by monitoring the first order decrease in ozone concentration as a function of excess alpha-terpineol. To better understand alpha-terpineol's gas-phase transformation in the indoor environment, the products of the alpha-terpineol + OH and alpha-terpineol + 03 reactions were also investigated. The positively identified alpha-terpineol/OH reaction products were acetone, ethanedial (glyoxal, HC(=O)C(=O)H), and 2-oxopropanal (methyl glyoxal, CH3C(=O)C(=O)H). The positively identified alpha-terpineol/O3 reaction product was 2-oxopropanal (methyl glyoxal, CH3C(=O)C(=O)H). The use of derivatizing agents O-(2,3,4,5,6-pentalfluorobenzyl)hydroxylamine (PFBHA) and N,O-bis(trimethylsilyl)trifluoroacetamide (BSTFA) clearly indicated that several other reaction products were formed. The elucidation of these other reaction products was facilitated by mass spectrometry of the derivatized reaction products coupled with plausible alpha-terpineol/OH and alpha-terpineol/O3 reaction mechanisms based on previously published volatile organic compound/ OH and volatile organic compound/O3 gas-phase reaction mechanisms.
Product Description
Supelco
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, for GC derivatization, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, ≥98%
Sigma-Aldrich
α-Terpineol, 90%, technical grade
Supelco
α-Terpineol, analytical standard
Supelco
α-Terpineol, analytical standard