MilliporeSigma
  • The use of thymidine analogs to improve the replication of an extra DNA base pair: a synthetic biological system.
The use of thymidine analogs to improve the replication of an extra DNA base pair: a synthetic biological system.
Nucleic acids research (2005-09-30)
A Michael Sismour, Steven A Benner
ABSTRACT
Synthetic biology based on a six-letter genetic alphabet that includes the two non-standard nucleobases isoguanine (isoG) and isocytosine (isoC), as well as the standard A, T, G and C, is known to suffer as a consequence of a minor tautomeric form of isoguanine that pairs with thymine, and therefore leads to infidelity during repeated cycles of the PCR. Reported here is a solution to this problem. The solution replaces thymidine triphosphate by 2-thiothymidine triphosphate (2-thioTTP). Because of the bulk and hydrogen bonding properties of the thione unit in 2-thioT, 2-thioT does not mispair effectively with the minor tautomer of isoG. To test whether this might allow PCR amplification of a six-letter artificially expanded genetic information system, we examined the relative rates of misincorporation of 2-thioTTP and TTP opposite isoG using affinity electrophoresis. The concentrations of isoCTP and 2-thioTTP were optimal to best support PCR amplification using thermostable polymerases of a six-letter alphabet that includes the isoC-isoG pair. The fidelity-per-round of amplification was found to be approximately 98% in trial PCRs with this six-letter DNA alphabet. The analogous PCR employing TTP had a fidelity-per-round of only approximately 93%. Thus, the A, 2-thioT, G, C, isoC, isoG alphabet is an artificial genetic system capable of Darwinian evolution.