MilliporeSigma
Continuous microbial fuel cells convert carbohydrates to electricity.
Water science and technology : a journal of the International Association on Water Pollution Research (2005-09-27)
I Rabaey, W Ossieur, M Verhaege, W Verstraete
ABSTRACT
Microbial fuel cells which are operated in continuous mode are more suitable for practical applications than fed batch ones. Three influent types containing carbohydrates were tested, i.e. a glucose medium, a plant extract and artificial wastewater. The anode reactor compartment yielding the best results was a packed bed reactor containing graphite granules. While in non-mediated batch systems power outputs up to 479 W m(-3) of anode compartment could be attained; in continuous mode the power outputs were limited to 49W m(-3). Cyclic voltammetry was performed to determine the potential of the in-situ synthesized bacterial redox mediators. Addition of mediators with a potential similar to the bacterial potential did not significantly alter the MFC power output, indicating a limited influence of soluble mediators for continuous microbial fuel cells. Maximum coulombic and energy conversion efficiencies were, for the continuous microbial fuel cell operating on plant extract at a loading rate of 1 kg COD m(-3) of anode compartment per day, 50.3% and 26.0% respectively.
Sigma-Aldrich
New Methylene Blue N, Dye content ~70 %