MilliporeSigma
  • Synthesis and studies on antioxidants: ethoxyquin (EQ) and its derivatives.
Synthesis and studies on antioxidants: ethoxyquin (EQ) and its derivatives.
Acta poloniae pharmaceutica (2005-09-16)
Alina Błaszczyk, Janusz Skolimowski
ABSTRACT
In our study ethoxyquin (EQ) and its two complexes with flavonoids were obtained from ethoxyquin (1,2-dihydro-6-ethoxy-2,2,4-trimethylquinoline, EQ) and quercetin (EQ-Q, 1:1) or rutin (EQ-R, 1:1). Cytotoxicity of the tested compounds was studied using the trypan blue exclusion method and the properties of the studied compounds were also analyzed with the TUNEL method evaluating their ability to induce apoptosis. It was shown that EQ induced apoptosis in cultured human lymphocytes, especially at 0.25 and 0.5 mM concentrations. The same effects were also observed after the incubation of lymphocytes with EQ-Q and EQ-R, but the numbers of apoptotic cells observed were lower than for EQ.
Sigma-Aldrich
Ethoxyquin, ≥75% (capillary GC)
Supelco
Ethoxyquin, PESTANAL®, analytical standard