MilliporeSigma
  • Biotransformation of racemic diisophorone by Cephalosporium aphidicola and Neurospora crassa.
Biotransformation of racemic diisophorone by Cephalosporium aphidicola and Neurospora crassa.
Biotechnology letters (2005-09-01)
Ismail Kiran, Tamer Akar, Asli Gorgulu, Cavit Kazaz
ABSTRACT
Racemic diisophorone (500 mg) was converted by Cephalosporium aphidicola and Neurospora crassa over 10 days at 25 degrees C to 8beta-hydroxydiisophorone in yields of 10% (52 mg) and 20% (103 mg), respectively. The structure was established by IR, specific rotation, mass spectral, 1D and 2D-NMR studies.
Sigma-Aldrich
Isophorone, 97%
Sigma-Aldrich
Isophorone, ≥97%, FG