MilliporeSigma
  • Inhibitory effects of Celecoxib and Sc-58125 on proliferation of human carcinoma of larynx Hep-2 in vitro.
Inhibitory effects of Celecoxib and Sc-58125 on proliferation of human carcinoma of larynx Hep-2 in vitro.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban (2005-08-25)
Juan Ding, Qing Chang, Shusheng Gong
ABSTRACT
The inhibitory effects of two kinds of selective cyclooxygenase-2 inhibitors on the proliferation of human carcinoma of larynx Hep-2 in vitro and their corresponding mechanisms were investigated. Hep-2 cells were cultured with two kinds of selective cyclooxygenase-2 inhibitors (Sc-58125 and Celecoxib) at various concentrations for 24 h. Morphological changes were observed under the phase microscopy and the growth suppression was detected by using MTT colorimetric assay. Apoptotic DNA fragments were observed by agarose gel electrophoresis, and the cell cycle and apoptotic rate were detected by flow cytometry (FCM) respectively. Hep-2 cells became rounded and detached from the culture dish after being treated with Celecoxib for 24 h, however, they remained morphologically unchanged with Sc-58125. Sc-58125 could increase G2 phase cells, whereas, Celecoxib rose G1 phase cells. Both of the two effects were dose-dependent. Moreover, the Hep-2 cells cultured with 50 micromol/L and 100 micromol/L Celecoxib showed obvious apoptosis, with the nuclear DNA of cells exhibiting characteristic DNA ladder. So Sc-58125 could inhibit the proliferation of Hep-2 cells by altering the G2 phase cells. However, Celecoxib had the same effect by changing the G1 phase cells and inducing apoptosis at higher concentration.
Sigma-Aldrich
SC-58125, ≥98% (HPLC)