MilliporeSigma
  • Detection of mercaptopyridines and mercaptopyrimidines in planar chromatography with iodine-azide reaction as a detection system.
Detection of mercaptopyridines and mercaptopyrimidines in planar chromatography with iodine-azide reaction as a detection system.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2005-08-16)
Robert Zakrzewski, Witold Ciesielski
ABSTRACT
Reaction between iodine and azide ion induced by mercaptopyridines and mercaptopyrimidines was utilized as a detection system in TLC and HPTLC. The developed plates were sprayed with a freshly prepared mixtures of sodium azide and starch solution adjusted to pH 5.5, and exposed to iodine vapour. The spots became visible as white spots on violet-grey background. The iodine-azide detection system has been proved to be the most favourable and enabled to detect quantities per spot in the range of 1-20 pmol (HPTLC) and 1-60 pmol (TLC). The iodine-azide tests were compared with other visualizing techniques commonly used in planar chromatography (iodine vapour and UV254). The developed method was applied to detection of thiopental in biological samples.
Sigma-Aldrich
4-Mercaptopyridine, 95%