MilliporeSigma
  • Immunohistochemical staining of epoxy resin sections of peripheral nerve.
Immunohistochemical staining of epoxy resin sections of peripheral nerve.
Applied immunohistochemistry & molecular morphology : AIMM (2005-08-06)
Zhao Cai, Jim Manavis, Kathy Cash, Philip D Thompson, Peter C Blumbergs
ABSTRACT
The authors describe a simple and cost-effective method of immunostaining semithin epoxy resin sections of peripheral nerve for light microscopy with antibodies to myelin protein zero, peripheral myelin protein 22, myelin basic protein, and neurofilament protein 200 using a combined technique of surface etching with sodium ethoxide and heat-induced antigen retrieval.
Sigma-Aldrich
Sodium ethoxide solution, 21 wt. % in ethanol
Sigma-Aldrich
Sodium ethoxide, technical, ≥95% (T)
Sigma-Aldrich
Sodium ethoxide, 95%