MilliporeSigma
Genetic analysis of reproduction in the Buryat populations.
Journal of physiological anthropology and applied human science (2005-08-05)
Nailija Khajievna Spitsyna, Victor Alexeevich Spitsyn, Vasilii Eugenievich Deryabin
ABSTRACT
A complex anthropological survey based on population-genetic methods and a study of a wide spectrum of genetic systems (43 alleles from 17 independent loci) was undertaken among 450 Buryat women of post-reproductive age. The results obtained showed the influence of particular genetic markers and their complex on the formation of peculiarities in the reproduction structure of the Buryat population.A sharp increase in phenotype GC 2-2 frequency and the corresponding GC*2 allele of the group-specific component (GC) was established for women groups with burdened obstetric records. These groups are characterized also by a considerable decrease in the observed geterozygosity (Ho) as compared to its expected value (He). Samples including women with multiple pregnancies in the recorded obstetric anamnesis are characterized by a significant increase in the frequency of the rare alleles TF*C3 of the transferrin system and those of PI*Z belonging to the proteinase inhibitor system (a1-antitrypsin) as compared to the control group. The results obtained widened current knowledge about the influence of genetic and environmental components on reproduction processes in human populations.
Sigma-Aldrich
DL-3-Aminoisobutyric acid, 98%