MilliporeSigma
  • [The effect of preparation method on the quality of shen-mai injection].
[The effect of preparation method on the quality of shen-mai injection].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2005-08-04)
Yong-jiang Wu, Qin-min Cui, Yi-yu Cheng
ABSTRACT
To investigate the effect of different preparation method on the quality of Shen-mai injection. The Shen-mai injection samples were prepared using three different methods. Fingerprints of Shen-mai extracts red ginseng, and its intermediates were obtained using an HPLC analytical procedure. The contents of ginsenoside Rg1, Rc and Rb1, and the gross saponins of Shen-mai extract were quantitatively mensured with HPLC procedures. There was significant difference in fingerprints and chemical contents of the injections prepared by the three different methods. The quality of Shen-mai injection was greatly influenced by the preparation method. HPLC fingerprinting method can be applied for the determination of the Shen-mai preparations.
Sigma-Aldrich
Ginsenoside-Rc from Panax ginseng (Korean ginseng) root, triterpenoid saponin, ≥98% (HPLC)