MilliporeSigma
A scalable route to trisubstituted (E)-vinyl bromides.
Organic letters (2005-07-29)
Cheon-Gyu Cho, Won-Suk Kim, Amos B Smith
ABSTRACT
An effective, readily scalable two-step synthesis of trisubstituted (E)-vinyl bromides involving bromination of alpha,beta-unsaturated lactones followed by hydrolytic fragmentation has been developed. Several trisubstituted (E)-vinyl bromides, including multigram quantities of (+)-(E)-4-bromo-2-methyl-3-pentenol, a synthetic intermediate required for the C(8)-C(11) moieties of (+)-tedanolide (1) and (+)-13-deoxytedanolide (2), illustrate the utility of this protocol. [reaction: see text]
Sigma-Aldrich
Vinyl bromide solution, 1.0 M in THF