MilliporeSigma
  • Chromatographic resolution of glucosidic compounds, ginsenosides on polyethersulphone membrane, and its application to the quantitative immunoassay for ginseng saponins.
Chromatographic resolution of glucosidic compounds, ginsenosides on polyethersulphone membrane, and its application to the quantitative immunoassay for ginseng saponins.
Glycobiology (2005-06-24)
Osamu Morinaga, Noriko Fukuda, Hiroyuki Tanaka, Yukihiro Shoyama
ABSTRACT
A method has been devised for the chromatographic resolution of glucosidic compounds, ginseng saponins, on polyethersulphone (PES) membrane. The method results in good resolution and quantitative immunoassay for ginsenoside Rb1 (G-Rb1), G-Rc, and G-Rd in crude extracts of various ginsengs. The newly established method is simpler and applies for quantitative analysis. Ginsenosides developed by acetonitrile-water-acetic acid solvent system on a PES membrane were directly treated with a NaIO4 solution followed by bovine serum albumin (BSA), resulting in a ginsenoside-BSA conjugate on a PES membrane. Anti-G-Rb1 monoclonal antibody (MAb) was bound, and then a second antibody labeled with peroxidase directed against the first antibody. Finally a substrate reacted to the enzyme and gave staining. The stained membrane was scanned, and spots were analyzed quantitatively using NIH Image software. At least 62.5 ng of G-Rb1, G-Rc, and G-Rd were clearly detectable individually. Three ginsenosides can be analyzed quantitatively between 0.125 and 2.0 microg.
Sigma-Aldrich
Ginsenoside-Rc from Panax ginseng (Korean ginseng) root, triterpenoid saponin, ≥98% (HPLC)