MilliporeSigma
  • Authenticity assessment of gamma- and delta-decalactone from prunus fruits by gas chromatography combustion/pyrolysis isotope ratio mass spectrometry (GC-C/P-IRMS).
Authenticity assessment of gamma- and delta-decalactone from prunus fruits by gas chromatography combustion/pyrolysis isotope ratio mass spectrometry (GC-C/P-IRMS).
Journal of agricultural and food chemistry (2005-06-23)
Hirotoshi Tamura, Markus Appel, Elke Richling, Peter Schreier
ABSTRACT
Authenticity assessment of gamma-decalactone (1) and delta-decalactone (2) from peach (Prunus persica var. persica), apricot (Prunus armeniaca), and nectarine (Prunus persica var. nectarina) was performed using gas chromatography-isotope ratio mass spectrometry (GC-IRMS) in the combustion (C) and pyrolysis (P) mode. In addition, commercially available synthetic (nature-identical) 1 and 2 as well as biotechnologically produced samples (declared to be "natural") were characterized by their delta(2)H(V)(-)(SMOW) and delta(13)C(V)(-)(PDB) values. For the Prunus fruits under study, rather narrow ranges of delta(13)C(V)(-)(PDB) and delta(2)H(V)(-)(SMOW) data of 1, varying from - 34.6 per thousand to - 38.4 per thousand and -160 per thousand to -206 per thousand, respectively, were obtained. Synthetic references of 1 showed delta(13)C(V)(-)(PDB) and delta(2)H(V)(-)(SMOW) data ranging from -27.4 per thousand to -28.3 per thousand and -151 per thousand to -184 per thousand, respectively. Samples of 1 declared to be "natural" exhibited ranges from -28.1 per thousand to -29.2 per thousand and -192 per thousand to -286 per thousand for delta(13)C(V)(-)(PDB) and delta(2)H(V)(-)(SMOW), respectively. For 2 from peach, apricot, and nectarine, delta(13)C(V)(-)(PDB) values ranging from -34.0 per thousand to -37.9 per thousand were determined; the delta(2)H(V)(-)(SMOW) values ranged from -171 per thousand to -228 per thousand. The delta(13)C(V)(-)(PDB) and delta(2)H(V)(-)(SMOW) data for synthetic 2 were -28.2 per thousand and -171 per thousand, respectively, that is, similar to those of 2 from "natural" origin, ranging from -27.7 per thousand to -30.1 per thousand and -185 per thousand to -230 per thousand for delta(13)C(V)(-)(PDB) and delta(2)H(V)(-)(SMOW), respectively. GC-C/P-IRMS allowed clear-cut analytical differentiation of the synthetic and "ex-plant" origin of 1 and 2, whereas narrow ranges of delta(13)C(V)(-)(PDB) and delta(2)H(V)(-)(SMOW) data were found for samples of synthetic and "natural" origin.
Sigma-Aldrich
γ-Decalactone, ≥98%, FG, FCC
Sigma-Aldrich
γ-Decanolactone, 98%