MilliporeSigma
  • Residues of the lampricides 3-trifluoromethyl-4-nitrophenol and niclosamide in muscle tissue of rainbow trout.
Residues of the lampricides 3-trifluoromethyl-4-nitrophenol and niclosamide in muscle tissue of rainbow trout.
Journal of agricultural and food chemistry (2005-06-23)
Terrance D Hubert, Jeffry A Bernardy, Chue Vue, Verdel K Dawson, Michael A Boogaard, Theresa M Schreier, William H Gingerich
ABSTRACT
Rainbow trout (Oncorhyncus mykiss) were exposed to the (14)C-labeled lampricide 3-trifluoromethyl-4-nitrophenol (TFM) (2.1 mg/L) or niclosamide (0.055 mg/L) in an aerated static water bath for 24 h. Fish were sacrificed immediately after exposure. Subsamples of skin-on muscle tissue were analyzed for residues of the lampricides. The primary residues in muscle tissue from fish exposed to TFM were parent TFM (1.08 +/- 0.82 nmol/g) and TFM-glucuronide (0.44 +/- 0.24 nmol/g). Muscle tissue from fish exposed to niclosamide contained niclosamide (1.42 +/- 0.51 nmol/g), niclosamide-glucuronide (0.0644 +/- 0.0276 nmol/g), and a metabolite not previously reported, niclosamide sulfate ester (1.12 +/- 0.33 nmol/g).
Sigma-Aldrich
4-Nitro-3-(trifluoromethyl)phenol, 99% (GC)