MilliporeSigma
Double carbonylation of zirconocene-alkyne complexes.
Chemical communications (Cambridge, England) (2005-05-12)
Shizue Mito, Tamotsu Takahashi
ABSTRACT
Zirconocene-alkyne complexes prepared from Cp2ZrBu2, phosphines and alkynes reacted with CO to give double carbonylation products, 4-hydroxycyclobuten-1-one derivatives after hydrolysis.
Sigma-Aldrich
Bis(cyclopentadienyl)zirconium(IV) dichloride, ≥98%