MilliporeSigma
  • Fluorescent sensing and selective Pb(II) extraction by a dansylamide ion-exchanger.
Fluorescent sensing and selective Pb(II) extraction by a dansylamide ion-exchanger.
Journal of the American Chemical Society (2005-05-05)
Konstantinos Kavallieratos, Jay M Rosenberg, Wei-Zhong Chen, Tong Ren
ABSTRACT
The (bis)dansylated sulfonamide 1,2-C6H4(NHSO2C10H6-5-N(CH3)2)2 (1) extracted Pb(II) selectively from water into 1,2-dichloroethane via an ion-exchange mechanism and showed fluorescence quenching upon Pb(II) extraction. The distribution ratios for metal extraction (determined by ICP-MS) for Pb(II) were 133-1410 times higher than those for other metal cations [Co(II), Ni(II), Cu(II), Zn(II), and Cd(II)] under identical conditions. Fluorescence quenching was observed upon Pb(II) extraction, which was dependent on Pb(II) concentration. The monodansylated control, C6H5NHSO2C10H6-5-N(CH3)2 (2), showed neither extraction nor quenching, indicating that the fluorescence effects are a direct result of Pb coordination to 1. The observed selectivity for Pb(II) is ascribed to the formation of a low-coordinate binary Pb(II)-Sulfonamido complex in the organic phase.
Sigma-Aldrich
5-(Dimethylamino)-1-naphthalenesulfonamide, 99%