MilliporeSigma
  • Increasing the accuracy of determination of nc/nH ratios by gas chromatography-atomic emission detection.
Increasing the accuracy of determination of nc/nH ratios by gas chromatography-atomic emission detection.
Journal of chromatography. A (2005-05-04)
Elena S Chernetsova, Alexander I Revelsky, Dupont Durst, Tim G Sobolevsky, Igor A Revelsky
ABSTRACT
The influence of oxygen content in helium on the accuracy of nc/nH ratio determination for model mixtures of aliphatic and polyaromatic hydrocarbons and polychlorinated biphenyls was studied. The best accuracy was achieved at the oxygen content ca. 9%, which was the maximal possible oxygen content in helium for this GC-atomic emission detection (helium flow rate was 25 ml min(-1)). Using the maximal oxygen flow in plasma the nC/nH ratio determination accuracy improvement was accompanied by 10-fold increase in detection limit.
Product Description
Supelco
Tridecane, analytical standard
Sigma-Aldrich
Tridecane, ≥99%