MilliporeSigma
  • 1H and 13C NMR identification of unexpected 3,4-dihydroquinoxalines in the syntheses of 1,5-benzodiazepine derivatives.
1H and 13C NMR identification of unexpected 3,4-dihydroquinoxalines in the syntheses of 1,5-benzodiazepine derivatives.
Magnetic resonance in chemistry : MRC (2005-04-30)
E M Tjiou, E G Lhoussaine, D Virieux, A Fruchier
ABSTRACT
The reaction between o-phenylenediamines, dehydroacetic acid and aromatic aldehydes is shown to give not only the expected 1,5-benzodiazepine derivative but also a 3,4-dihydroquinoxaline, the structure of which was determined by its 1H and 13C 1D and 2D NMR spectra.
Sigma-Aldrich
Dehydroacetic acid, ≥98.0% (T)