MilliporeSigma
Textile dye decolorization using cyanobacteria.
Biotechnology letters (2005-04-19)
Amit Parikh, Datta Madamwar
ABSTRACT
Cyanobacterial cultures isolated from sites polluted by industrial textile effluents were screened for their ability to decolorize cyclic azo dyes. Gloeocapsa pleurocapsoides and Phormidium ceylanicum decolorized Acid Red 97 and FF Sky Blue dyes by more than 80% after 26 days. Chroococcus minutus was the only culture which decolorized Amido Black 10B (55%). Chlorophyll a synthesis in all cultures was strongly inhibited by the dyes. Visible spectroscopy and TLC confirmed that color removal was due to degradation of the dyes.
Sigma-Aldrich
Amido Black Staining Solution 2X, electrophoresis reagent
Sigma-Aldrich
Naphthol Blue Black, Dye content ~50 %
Sigma-Aldrich
Naphthol Blue Black, BioReagent, suitable for electrophoresis
Sigma-Aldrich
Naphthol Blue Black, Dye content 80 %