MilliporeSigma
  • An approach to investigating the importance of high potency polycyclic aromatic hydrocarbons (PAHs) in the induction of lung cancer by air pollution.
An approach to investigating the importance of high potency polycyclic aromatic hydrocarbons (PAHs) in the induction of lung cancer by air pollution.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2005-04-19)
K B Okona-Mensah, J Battershill, A Boobis, R Fielder
ABSTRACT
Evidence suggests that people living in urban areas have an increased risk of lung cancer due to higher levels of air pollution in these areas. Benzo[a]pyrene (B[a]P) is currently used as the main indicator of carcinogenic polycyclic aromatic hydrocarbons (PAHs) in air pollution, but there is concern that B[a]P may not be the ideal surrogate of choice for PAH mixtures since higher potency PAHs have recently been identified which could potentially contribute more and variably to the overall carcinogenicity. Dibenzo[a,h]anthracene (DBA) and dibenzo[a,l]pyrene (DB[a,l]P) are estimated to have carcinogenic potencies 10 or more times greater than B[a]P but data on their presence and formation in the environment are limited. Several occupational and environmental PAH biomonitoring studies are reviewed here, with particular focus on the specific exposure groups, study design, sample tissue, in particular the use of nasal tissues, and biomarkers used in each study. Consideration of these data is then used to propose a novel biomonitoring approach to evaluate exposure, uptake and the role of high potency PAHs in air pollution-related lung cancer. This is based upon an occupational study examining specific DNA adducts for DBA and DB[a,l]P in nasal cells to evaluate the extent to which these high potency PAHs might contribute to the increased risk of developing lung cancer from air pollution.
Product Description
Dibenz[a,h]anthracene, BCR®, certified reference material
Supelco
Dibenz[a,h]anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland